Learn More
ATCC Corynebacterium riegelii
Catalog No. 502380411
Encompass
Corynebacterium riegelii strain CIP 105310 is a whole-genome sequenced bacterial type strain that was isolated from a patient with a urinary tract infection.
Catalog No. 50-238-0411 Supplier ATCC Supplier No. 700782
Corynebacterium riegelii strain CIP 105310 is a whole-genome sequenced bacterial type strain that was isolated from a patient with a urinary tract infection.