Learn More
ATCC Bacteroides fragilis; genomic DNA
Catalog No. 502381875
Encompass
Genomic DNA from Bacteroides fragilis strain VPI 2553 (ATCC 25285). This whole-genome sequenced product can be used in PCR and other molecular biology applications.
Catalog No. 50-238-1875 Supplier ATCC Supplier No. 25285D5
Genomic DNA from Bacteroides fragilis strain VPI 2553 (ATCC 25285). This whole-genome sequenced product can be used in PCR and other molecular biology applications.