Learn More
New England Biolabs, Inc. Bacteroides Heparinase I – 240 units
SDP
Catalog No. 50591171
Encompass
Bacteroides Heparinase I, also called Heparin Lyase I, is active on heparin and the highly sulfated domains of heparan sulfate. The reaction yields oligosaccharide products containing unsaturated uronic acids which can be detected by UV spectroscopy at 232 nm.
Catalog No. 50-591-171 Supplier New England Biolabs, Inc. Supplier No. P0735S
Bacteroides Heparinase I, also called Heparin Lyase I, is active on heparin and the highly sulfated domains of heparan sulfate. The reaction yields oligosaccharide products containing unsaturated uronic acids which can be detected by UV spectroscopy at 232 nm.