Learn More
New England Biolabs, Inc. Bacteroides Heparinase III – 14 units
SDP
Catalog No. 50591175
Encompass
Bacteroides Heparinase III, also called Heparin Lyase III, is active on both heparin and heparan sulfate. Bacteroides Heparinase III can cleave the glycosidic bond between hexosamines and either iduronic acid or glucuronic acid residues. It is active in the presence of 6-sulfation. The reaction yields oligosaccharide products containing unsaturated uronic acids, which can be detected by UV spectroscopy at 232 nm
Catalog No. 50-591-175 Supplier New England Biolabs, Inc. Supplier No. P0737S
Bacteroides Heparinase III, also called Heparin Lyase III, is active on both heparin and heparan sulfate. Bacteroides Heparinase III can cleave the glycosidic bond between hexosamines and either iduronic acid or glucuronic acid residues. It is active in the presence of 6-sulfation. The reaction yields oligosaccharide products containing unsaturated uronic acids, which can be detected by UV spectroscopy at 232 nm