Learn More
ATCC Azotobacter vinelandii; Strain DJ
Catalog No. 502381238
Encompass
Azotobacter vinelandii strain DJ is a whole-genome sequenced bacterium. This strain is grown aerobically on Azobacter supplement media.
Catalog No. 50-238-1238 Supplier ATCC Supplier No. BAA1303
Azotobacter vinelandii strain DJ is a whole-genome sequenced bacterium. This strain is grown aerobically on Azobacter supplement media.