Learn More
ATCC Bacteroides ovatus
Catalog No. 502382413
Encompass
Bacteroides ovatus strain [NCTC 11153] is a whole-genome sequenced bacterial type strain. This bacterial culture can be used as a quality control strain for API and BBL products or as a control strain for anaerobe identification.
Catalog No. 50-238-2413 Supplier ATCC Supplier No. 8483
Add to Cart
Add to Cart
Bacteroides ovatus strain [NCTC 11153] is a whole-genome sequenced bacterial type strain. This bacterial culture can be used as a quality control strain for API and BBL products or as a control strain for anaerobe identification.