Learn More
ATCC Bacteroides thetaiotaomicron
Supplier:  ATCC 29741
Encompass
Bacteroides thetaiotaomicron strain WAL 2926 [CIP 104207] was isolated from a perforated human appendix. This whole-genome sequenced strain is used in susceptibility testing, quality control of cefoperazone-sulbactam, and as a quality control strain for CLSI and for BBL and Micro-Media products.
Catalog No. 50-238-2904