Learn More
Fisherbrand™ Bacti-Loop™ Tilt Sterilizer: Heating Element
Catalog No. 22630007 Shop All Fisherbrand Products
Change view
Click to view available options
Model:
B1001-XL-HE
Catalog No. Model
22-630-007 B1001-XL-HE
1 options
Catalog No. 22-630-007
Add to Cart
Add to Cart
Replacement heating element for the Fisherbrand™ Bacti-Loop™ Tilt Sterilizer. The Bact-Loop™ has an asbestos-free core element that uses infrared high temperatures 815™C (1500™F).
Ceramic heating element, for use with the Fisherbrand™ Bacti-Loop™ Tilt Sterilizer.  
Specifications
For Use With (Equipment) Fisherbrand Bacti-Loop Tilt Sterilizer (22-630-006)
Model B1001-XL-HE
Product Type Replacement Heating Element