Learn More
Fisherbrand™ Bacti-Loop™ Tilt Sterilizer
Catalog No. 22630005 Shop All Fisherbrand Products
Change view
Click to view available options
Model:
B1001
B1001-XL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model
22-630-005 B1001
22-630-006 B1001-XL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-630-005
Add to Cart
Edge
Add to Cart
Fisherbrand™ Bacti-Loop™ Tilt Sterilizer has an asbestos-free core element that uses infrared high temperatures 815™C (1500™F).
  • Complete sterilization in seconds without spatter
  • Ideal for platinum loops and needles
  • Tilt model for hands-free sterilization of self-standing vessels
  • Available as XL, with wide chamber for sterilizing larger vessels
  • Completely sterilizes microorganisms within 5 to 7 seconds.
  • Provides instant sterilization of platinum inoculating loops, needles, glass tube/pipette mouths, and various metal and borosilicate glass instruments used in the laboratory.
  • Ceramic heating element is safely enclosed in a perforated
    stainless steel guard chamber, protecting users from accidental
    contact with the extreme temperatures of the core area.
  • Compact, convenient and can be used in anaerobic and aerobic chambers.
Specifications
Model B1001
Product Type Sterilizer