Learn More
Research Products International Corp Bacti Zapper Infrared Sterilizer
SDP
Catalog No. 501977212
Encompass
Bacti Zapper Infrared Sterilizer features an asbestos free core element with infrared heating that produces a temperature of 815° C (1500° F). Instantly sterilizes platinum inoculating loops, needles and a variety of metal and borosilicate glass instruments. The ceramic heating element is safety enclosed within a stainless steel chamber for maximum protection. Compact size allows unit to be used in anaerobic and aerobic chambers.
Catalog No. 50-197-7212 Supplier Research Products International Corp Supplier No. 248570
Bacti Zapper Infrared Sterilizer features an asbestos free core element with infrared heating that produces a temperature of 815° C (1500° F). Instantly sterilizes platinum inoculating loops, needles and a variety of metal and borosilicate glass instruments. The ceramic heating element is safety enclosed within a stainless steel chamber for maximum protection. Compact size allows unit to be used in anaerobic and aerobic chambers.