Learn More
Braintree Scientific Bacti Zapper Infrared Microsterilizer
SDP
Supplier:  Braintree Scientific B1000120V
Encompass
Infrared Microsterilizer BactiZapper™ has an asbestos-free core element that uses infrared heating to produce 815°C (1500°F). It completely sterilizes micro-organisms in 5-7 seconds. BactiZapper uses well-proven technology to provide instant sterilization of platinum inoculating loops, needles, glass tube pipette mouths and various metal and borosilicate glass instruments. Ceramic heating element is safely enclosed in a perforated stainless steel guard chamber, protecting users from accidental contact with extreme temperatures of the core area. It is extremely compact, convenient and can be used in anaerobic and aerobic chambers. Complete infrared sterilization in 5-7 seconds. Safe, convenient and no spatter. Ideal for platinum loops, needles and tube mouths. Specifications: Max temperature: 815° C /1500° F, Sterilization: 5-7 secs, Chamber diameter: .55"/14mm, Chamber length: 3.94"/100mm, External dimensions (W x D x H): 4" x 6" x 6.5"
Catalog No. 50-195-4255