Learn More
Thermo Scientific™ BactiSwab™ Gel Collection and Transport Swabs: Dual Swab Amies Clear
GSA_VA
Catalog No. R723015 Shop All Thermo Scientific Products
Change view
Click to view available options
Packaging:
50/Pk.
500/Pk.
Swab Configuration:
Dual Applicator
Single Applicator
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging Swab Configuration
R723015 50/Pk. Dual Applicator
R723005 50/Pk. Single Applicator
R723000 500/Pk. Single Applicator
R723010 500/Pk. Dual Applicator
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. R723015 Supplier Thermo Scientific™ Supplier No. R723015
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Thermo Scientific™ Remel BactiSwab Gel Amies Clear are supplied with double plastic shaft.
Thermo Scientific™ Remel BactiSwab Gel Amies Clear are supplied with double plastic shaft.
Rayon-tipped, plastic-shaft swab(s) are mounted in the cap of a translucent plastic tube with spiral chamber containing gel transport medium with or without charcoal.
  • Blue cap
  • Round-bottom tube for easy racking and seamless molding to ensure strength and integrity
  • Exclusive zip-seal packaging to promote optimum media performance
  • Color-coded cap with protective overskirt
  • Inert, non-toxic rayon swab tip for good sample retrieval and absorption
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Format Tube
Media Type Amies Clear
Swab Configuration Dual Applicator
Closure Color Blue
Description Amies Clear
Packaging 50/Pk.
Quantity 1 Tube/Envelope
Shaft Material Plastic