Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ BactiSwab™: Liquid Stuart Plastic Shaft

Catalog No. R723125 Shop All Thermo Scientific Products
Change view
Click to view available options
Swab Configuration:
Dual Applicator
Single Applicator
Closure Color:
Red
White
Quantity:
50 Pk.
500 Pk.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Swab Configuration Closure Color Quantity
R723125 Dual Applicator Red 50 Pk.
R723110 Single Applicator White 500 Pk.
R723120 Dual Applicator Red 500 Pk.
R723115 Single Applicator White 50 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. R723125 Supplier Thermo Scientific™ Supplier No. R723125
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Thermo Scientific™ Remel BactiSwab™ Liquid Stuart, Plastic Shaft is supplied with a single or double plastic shaft.

Rayon-tipped, plastic-shaft swab(s) are mounted in the cap of a translucent plastic tube with spiral chamber containing a sponge impregnated with liquid transport medium.
TRUSTED_SUSTAINABILITY

Specifications

Format Tube
Media Type Liquid Stuart
Swab Configuration Dual Applicator
Closure Color Red
Description Liquid Stuarts
For Use With (Application) Healthcare
Quantity 50 Pk.
Shaft Material Plastic
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.