Description
Thermo Scientific™ Remel BactiSwab contains a single plasti-shaft swab.
Rayon-tipped, plastic-shaft swabs are mounted in the cap of a translucent plastic tube. Inside the tube, unless otherwise noted in product description that it is ‘dry’, is a crushable ampule of Modified Stuart's Medium with a pledget that becomes moistened with the medium when ampule is crushed.
Ordering Alerts:
Each BactiSwab is supplied as one tube per envelope.
Specifications
Specifications
| Swab Configuration | Single Applicator |
| Description | Bacti-Swab; Single plastic-shaft swab |
| For Use With (Application) | Healthcare |
| Quantity | 100/Pk. |
| Shaft Material | Plastic |
| Tip Material | Rayon |