Learn More
Thomas Scientific Infared MicroSterilizer, 120V
SDP
Catalog No. 501460440
Encompass_Preferred
Complete sterilization of loops, and tube mouths Safe, convenient and no spatter Eliminates the need for an open flame Ideal for research laboratories The labForce MicroSterilizer is a research sterilizer that utilizes an asbestos-free core element that uses intense heating to produce 815°C (1500°F). It completely sterilizes in 5-7 seconds. The MicroSterilizer uses well-proven technology to provide instant sterilization of platinum inoculating loops, glass tube/pipette mouths and various metal & borosilicate glass instruments used in quality control and research. It also has a lower temperature setting (500°C) that can be used as a "standby" and extends usable life. The ceramic heating element is safely enclosed in a perforated stainless steel guard chamber, protecting users from accidental contact with the extreme temperatures of the core area. It is extremely compact, convenient and can be used in anaerobic and aerobic chambers.
Catalog No. 50-146-0440 Supplier Thomas Scientific Supplier No. 1154J15
Complete sterilization of loops, and tube mouths Safe, convenient and no spatter Eliminates the need for an open flame Ideal for research laboratories The labForce MicroSterilizer is a research sterilizer that utilizes an asbestos-free core element that uses intense heating to produce 815°C (1500°F). It completely sterilizes in 5-7 seconds. The MicroSterilizer uses well-proven technology to provide instant sterilization of platinum inoculating loops, glass tube/pipette mouths and various metal & borosilicate glass instruments used in quality control and research. It also has a lower temperature setting (500°C) that can be used as a "standby" and extends usable life. The ceramic heating element is safely enclosed in a perforated stainless steel guard chamber, protecting users from accidental contact with the extreme temperatures of the core area. It is extremely compact, convenient and can be used in anaerobic and aerobic chambers.