Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Bacto™ Casitone
GREENER_CHOICE

Catalog No. p-8843004
Change view
Click to view available options
Quantity:
500 g
10 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DF0259-17-9 500 g
22-591-0 10 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. DF0259-17-9 Supplier Gibco™ Supplier No. 225930
Only null left
Add to Cart
Add to Cart

An animal origin (AO) pancreatic enzymatic digest of casein.

  • An animal origin (AO) pancreatic enzymatic digest of casein similar to Difco Casein Digest
  • This supplement provides a nutritional profile based on casein
  • The specific enzymatic digest performed for this product results in a particularly high content of amino acids and peptides of varying sizes, making it a more nutritious hydrolysate
  • It has been successfully used for the production of Escherichia coli-based recombinant proteins
  • Store in cool, dry conditions in well-sealed containers at 2°C to 25°C
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO/DIS, 11014
Content And Storage Storage conditions: 2°C to 25°C. Store in cool, dry conditions in well-sealed containers.
Form Powder
Product Type Dehydrated Culture Media
Format Bottle
For Use With (Application) Production of Escherichia coli-based recombinant proteins
Product Line Bacto
Quantity 500 g
Source Animal origin, casein
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.