Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Bacto™ CD Supreme Fermentation Production Medium Plus, 1 L

Catalog No. A9378401
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A9378401 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A9378401 Supplier Gibco™ Supplier No. A9378401
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemically defined medium optimized for E. coli plasmid DNA and recombinant protein production.

Gibco Bacto CD Supreme Fermentation Production Medium (FPM) Plus is the next-generation, chemically defined formulation designed to support a wide range of Escherichia coli (E. coli) strains used in plasmid DNA and recombinant protein production. This hydrolysate-free medium enhances microbial culture performance in both batch and fed-batch fermentation processes, offering superior consistency and flexibility across manufacturing scales.

Features of Bacto CD Supreme FPM Plus include:

  • Consistency with a 100% chemically defined, hydrolysate-free formulation that minimizes lot-to-lot variability
  • Expanded strain compatibility to include E. coli strains such as DH10B, BL21(DE3), NEB™ Stable, Stbl2, Stbl3, JM109, and other auxotrophic variants
  • Simplified workflows using a one-part, ready-to-use formulation that eliminates manual media preparation
  • Easy adaptation to your process with format options and compatibility with filtration or autoclave sterilization methods
  • Reduced regulatory risk with a formulation free from raw materials of animal origin or materials produced using animal-derived components

Bacto CD Supreme FPM Plus is engineered to deliver reliable and reproducible growth performance across diverse E. coli host strains, including Thi-1 mutant and auxotrophic variants. Its chemically defined composition helps ensure optimal nutrient availability without the variability introduced by hydrolysates. The medium supports high-density cultures in both batch and fed-batch modes, enabling efficient plasmid DNA or recombinant protein yield.

For fed-batch processes Bacto CD Supreme FPM Plus can be combined with Bacto CD Supreme Feed (Cat. Nos. A9378201, A9378301, A9378302) to support increased biomass accumulation and yields

This one-part, easy-to-use formulation integrates smoothly into existing manufacturing workflows and is compatible with standard sterilization methods. It is produced without any materials of animal origin, minimizing regulatory complexity and eliminating TSE/BSE risk concerns.

Specifications

Concentration 1x
Content And Storage Store in the dark at +2°C to +8°C
Form Liquid
Manufacturing Origin United States
Product Type Culture Medium
Sterility Sterile
Target Organism Class E. coli
Test Type Microbial Cultivation
Formulation Animal Origin-free, Chemically-defined
For Use With (Application) Microbial Fermentation
Product Line Bacto CD Supreme
Quantity 1 L
Show More Show Less

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.