Learn More
Bioworld Bacto Peptone, 73049-73-7, 500g
Catalog No. 50254270
Encompass_Preferred
Used as an organic nitrogen source in microbiological media for culturing variety of bacteria and fungi. Enzymatic digest of animal protein, Working concentration: 50 mg/mL, Dehydrated Appearance: faint yellow to yellow-brown powder, Prepared Appearance: dark yellow solution, Soluble in water, Appearance Form : Solid, Appearance Color : Faint yellow to yellow-brown. 500g
Catalog No. 50-254-270 Supplier Bioworld Supplier No. 306200052
Used as an organic nitrogen source in microbiological media for culturing variety of bacteria and fungi. Enzymatic digest of animal protein, Working concentration: 50 mg/mL, Dehydrated Appearance: faint yellow to yellow-brown powder, Prepared Appearance: dark yellow solution, Soluble in water, Appearance Form : Solid, Appearance Color : Faint yellow to yellow-brown. 500g