Learn More
McKesson General Medical BactoShield CHG 4% Handwash, Antimicrobial Skin Cleanser, Non-Sterile, 32 oz
Catalog No. NC2401580
Encompass
BactoShield CHG 4% Handwash / Surgical Scrub is a non-sterile, antimicrobial skin cleanser that effectively cleans and prepares the skin, providing rapid and lasting antimicrobial action against a wide range of microorganisms. With a strength of 4% Chlorhexidine Gluconate in a 32 oz bottle, it is ideal for healthcare personnel handwashing, surgical scrubbing, and general skin cleaning. This product stands out due to its proven efficacy and convenient liquid form.
Catalog No. NC2401580 Supplier McKesson General Medical Supplier No. 415473
BactoShield CHG 4% Handwash / Surgical Scrub is a non-sterile, antimicrobial skin cleanser that effectively cleans and prepares the skin, providing rapid and lasting antimicrobial action against a wide range of microorganisms. With a strength of 4% Chlorhexidine Gluconate in a 32 oz bottle, it is ideal for healthcare personnel handwashing, surgical scrubbing, and general skin cleaning. This product stands out due to its proven efficacy and convenient liquid form.