Learn More
Molecular Research Center BACTOZOL KIT
SDP
Catalog No. NC9326164
Encompass
A kit for the isolation of bacterial DNA. Kit includes 10X Bactozol Enzyme Solution, Dilution Buffer and DNAzol sufficient for 125 isolations, each yielding up to 40 ug bacterial DNA.
Catalog No. NC9326164 Supplier Molecular Research Center Supplier No. BA 154
A kit for the isolation of bacterial DNA. Kit includes 10X Bactozol Enzyme Solution, Dilution Buffer and DNAzol sufficient for 125 isolations, each yielding up to 40 ug bacterial DNA.