Learn More
Cayman Chemical Bacterial Fatty Acid Methyl Ester Mixture 2, 1 mL
SDP
Catalog No. 502989155
Encompass_Preferred
A mixture of bacterial FAMEs; intended for use as a reference standard for the detection of these bacterial fatty acids in various sample types by GC-FID or GC-MS
Catalog No. 50-298-9155 Supplier Cayman Chemical Supplier No. 293731 ML
A mixture of bacterial FAMEs; intended for use as a reference standard for the detection of these bacterial fatty acids in various sample types by GC-FID or GC-MS