Learn More
Cayman Chemical Bacterial Fatty Acid Methyl Ester Mixture 1, 1 mL
SDP
Catalog No. 502988807
Encompass_Preferred
A mixture of bacterial FAMEs; intended for use as a reference standard for the detection of these saturated branched- and straight-chain fatty acids in various sample types by GC-FID or GC-MS
Catalog No. 50-298-8807 Supplier Cayman Chemical Supplier No. 293721 ML
A mixture of bacterial FAMEs; intended for use as a reference standard for the detection of these saturated branched- and straight-chain fatty acids in various sample types by GC-FID or GC-MS