Learn More
iQ Biosciences Bactrian Camel PBMCs, 50M cells/vial
Catalog No. NC3235564
Encompass_Preferred
High-quality, high-viability (> 80%) bactrian camel peripheral blood mononuclear cells (PBMCs) sourced responsibly from a vetted third party facility. Potential uses include, production of stable single-domain antibodies (nanobodies) for therapeutic and diagnostic applications, vaccine development, and autoimmune research. All orders come with an iQ Certificate of Analysis.
Catalog No. NC3235564 Supplier iQ Biosciences Supplier No. IQBCMPB150
Add to Cart
Add to Cart
High-quality, high-viability (> 80%) bactrian camel peripheral blood mononuclear cells (PBMCs) sourced responsibly from a vetted third party facility. Potential uses include, production of stable single-domain antibodies (nanobodies) for therapeutic and diagnostic applications, vaccine development, and autoimmune research. All orders come with an iQ Certificate of Analysis.