Learn More
Sheldon Manufacturing Inc BACTRONEZ Rolling Stand
SDP
Catalog No. NC1884669
Encompass_Preferred
BACTRONEZ Rolling Stand for use with the BACTRONEZ has the dimenstions of (W x D x H) 49.0” x 30” x 30”/1240 x 762 x 762 mm.
Catalog No. NC1884669 Supplier Sheldon Manufacturing Inc Supplier No. BACSTANDSM22
BACTRONEZ Rolling Stand for use with the BACTRONEZ has the dimenstions of (W x D x H) 49.0” x 30” x 30”/1240 x 762 x 762 mm.