Learn More
Aviva Systems Biology Corp BACULOQUANT ALL-IN-ONE KIT
SDP
Catalog No. NC1889208
Encompass
BaculoQUANT All-In-One Virus Extraction & Titration Kit (GWB-100602). • Titre accuracy comparable to plaque assay method• Less than one hour hands-on time• Virus DNA extraction and titration in under three hours• Compatible with any AcMNPV-based baculovirus system containing gp64 viral gene• Single step virus DNA extraction: just add lysis solution (no spin columns)• QPCR Primers/ Probe provided as a single reagent mix: just add water• Agilent high performance, ultra sensitive master mix reagent included• Plaque assay titred virus internal standards included for standard curve generation
Catalog No. NC1889208 Supplier Aviva Systems Biology Corp Supplier No. GWB100602
BaculoQUANT All-In-One Virus Extraction & Titration Kit (GWB-100602). • Titre accuracy comparable to plaque assay method• Less than one hour hands-on time• Virus DNA extraction and titration in under three hours• Compatible with any AcMNPV-based baculovirus system containing gp64 viral gene• Single step virus DNA extraction: just add lysis solution (no spin columns)• QPCR Primers/ Probe provided as a single reagent mix: just add water• Agilent high performance, ultra sensitive master mix reagent included• Plaque assay titred virus internal standards included for standard curve generation