Learn More
MEDNA SCIENTIFIC INC Baculovirus Particles (BVPs) - 100uL
Catalog No. NC1999901
Encompass
Empty baculovirus particles (BVPs) (empty viral capsids without genomes) can be used to measure polyspecificity of therapeutic antibody candidates in in vitro assays. This assesses the viability of an antibody as a therapy early on. They are provided as viral particles in phosphate buffered saline (PBS) at aconcentration of 1 x 1012-13 pfu/mL. 100 uL is enough to assay 4 x 96-well plates at a final dilution of 1:200. For more information, please visit mednabio.com. Shipped and stored frozen.
Catalog No. NC1999901 Supplier MEDNA SCIENTIFIC INC Supplier No. E3001
Empty baculovirus particles (BVPs) (empty viral capsids without genomes) can be used to measure polyspecificity of therapeutic antibody candidates in in vitro assays. This assesses the viability of an antibody as a therapy early on. They are provided as viral particles in phosphate buffered saline (PBS) at aconcentration of 1 x 1012-13 pfu/mL. 100 uL is enough to assay 4 x 96-well plates at a final dilution of 1:200. For more information, please visit mednabio.com. Shipped and stored frozen.