Learn More
Aviva Systems Biology Corp BAD Antibody - C-terminal region (ARP30325_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP30325_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against BAD. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human BAD. Alternate names: BBC2, BCL2L8ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-54156