Learn More
Enzo Life Sciences Bad polyclonal antibody (200µg)
SDP
Supplier:  Enzo Life Sciences ADIAAP020F
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to a portion of mouse Bad. Source: Purified from rabbit serum. Species reactivity: Mouse, Rat, Sheep, Rabbit, Porcine, Monkey, Hamster, Drosophila, Bovine. Applications: IHC, WB. Purity: Protein A affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol and 0.09% sodium azide.
Catalog No. 50-200-7147