Learn More
Apexbio Technology LLC BADGE 1675-54-3 10mg
SDP
Catalog No. 502240633
Encompass_Preferred
BADGE (CAS nan) is a small-molecule antagonist targeting peroxisome proliferator-activated receptor gamma (PPAR ) It is designed to inhibit PPAR activity thereby modulating receptor-mediated signaling pathways involved in immune regulation BADGE exerts its biological activity primarily by antagonizing PPAR resulting in altered immune responses such as increased histamine secretion from immune cells and changes in cell-mediated immune mechanisms Based on these pharmacological properties BADGE holds research potential in investigating immune-related biological pathways allergic reactions and chemical sensitization mechanisms at the molecular and cellular levels
This item is not returnable. View return policy
Catalog No. 50-224-0633 Supplier Apexbio Technology LLC Supplier No. B669110
BADGE (CAS nan) is a small-molecule antagonist targeting peroxisome proliferator-activated receptor gamma (PPAR ) It is designed to inhibit PPAR activity thereby modulating receptor-mediated signaling pathways involved in immune regulation BADGE exerts its biological activity primarily by antagonizing PPAR resulting in altered immune responses such as increased histamine secretion from immune cells and changes in cell-mediated immune mechanisms Based on these pharmacological properties BADGE holds research potential in investigating immune-related biological pathways allergic reactions and chemical sensitization mechanisms at the molecular and cellular levels