Learn More
Enzo Life Sciences BADGE (25g). CAS: 1675-54-3
SDP
Supplier:  Enzo Life Sciences ALX270353G025
Encompass_Preferred
Antagonist of PPARγ. Binds to PPARγ with an apparent Kd of 100µM. Alternative name: Bisphenol A diglycidyl ether, 2,2'-[(1-Methylethylidene)bis(4,1-phenyleneoxymethylene)]bis-oxirane. Formula: C21H24O4. MW: 340.4. Purity: ≥85%. Appearance: Colorless viscous liquid. Solubility: Miscible in organic solvents as 100% ethanol, DMSO or dimethyl formamide; insoluble in PBS, pH 7.2. Long Term Storage: -20°C.
Catalog No. 50-200-8504