Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DR Instruments Badge Holder Retractable Reel
SDPSCI_ED

Catalog No. S20678
Change view
Click to view available options
Color:
Black
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
S20678 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options

California Proposition 65 WARNING: Cancer and Reproductive Harm

Catalog No. S20678 Supplier DR Instruments Supplier No. DRRTBH1
Add to Cart
Add to Cart

California Proposition 65 WARNING: Cancer and Reproductive Harm

Heavy duty retractable reel with belt clip and plastic ID holder.

  • Retractable 24 in. cord securely holds up to five keys, slot-punched photo ID or small tools.
  • Withstands everyday bumps and scuffs.
  • Includes a plastic horizontal I.D. holder to easily secure I.D. to badge reel.

Specifications

Product Type Badge Holder
Includes I.D. Holder
Color Black
For Use With (Application) Hold keys, photo ID or small tools
Diameter (English) 1.5 in.
Diameter (Metric) 38.1 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.