Learn More
New England Biolabs, Inc. BaeGI – 500 units
SDP
Catalog No. 50995284
Encompass
A restriction endonuclease that recognizes the sequence G_KGCM^C.
Catalog No. 50-995-284 Supplier New England Biolabs, Inc. Supplier No. R0708S
A restriction endonuclease that recognizes the sequence G_KGCM^C.