Learn More
New England Biolabs, Inc. BaeI – 250 units
SDP
Catalog No. 50812348
Encompass
A restriction endonuclease that recognizes the sequence _NNNNN^NNNNNNNNNNACNNNNGTAYCNNNNNNN_NNNNN^.
Catalog No. 50-812-348 Supplier New England Biolabs, Inc. Supplier No. R0613S
A restriction endonuclease that recognizes the sequence _NNNNN^NNNNNNNNNNACNNNNGTAYCNNNNNNN_NNNNN^.