Learn More
Cell Signaling Technology BAF Complex Antibody Sampler Kit II, Unconjugated, 1 Kit
SDP
Catalog No. 502732794
Encompass
The BAF Complex Antibody Sampler Kit II provides an economical means of detecting levels of various BAF complex components. The kit contains enough primary antibodies to perform at least two western blot experiments.
  • Includes seven primary antibodies for investigating BAF complex components.
  • Supplied in 10 mM sodium HEPES (pH 7.5), 150 mM NaCl, 100 μg/mL BSA, 50% glycerol and less than 0.02% sodium azide.
  • Validated for multiple applications including western blotting, immunoprecipitation, and chromatin IP-seq.
  • Includes secondary anti-rabbit IgG, HRP-linked antibody for western blot detection.
Catalog No. 50-273-2794 Supplier Cell Signaling Technology Supplier No. 58214T
The BAF Complex Antibody Sampler Kit II provides an economical means of detecting levels of various BAF complex components. The kit contains enough primary antibodies to perform at least two western blot experiments.
  • Includes seven primary antibodies for investigating BAF complex components.
  • Supplied in 10 mM sodium HEPES (pH 7.5), 150 mM NaCl, 100 μg/mL BSA, 50% glycerol and less than 0.02% sodium azide.
  • Validated for multiple applications including western blotting, immunoprecipitation, and chromatin IP-seq.
  • Includes secondary anti-rabbit IgG, HRP-linked antibody for western blot detection.