Learn More
FIREBIRD OPTICS Barium Fluoride (BaF2) Circular Optical Window- 13x2mm
SDP
Catalog No. NC1953391
Encompass_Preferred
Barium Fluoride (BaF2) Circular Optical Window- 13x2mm • Wavelength range: 200nm-12µm •Surface quality: 60/40 • Clear aperture: 90% •Diameter tolerance: ±0.1mm •Thickness tolerance: ±0.2mm
Catalog No. NC1953391 Supplier FIREBIRD OPTICS Supplier No. BAF213X2
Barium Fluoride (BaF2) Circular Optical Window- 13x2mm • Wavelength range: 200nm-12µm •Surface quality: 60/40 • Clear aperture: 90% •Diameter tolerance: ±0.1mm •Thickness tolerance: ±0.2mm