Learn More
Medchemexpress LLC HY-50868 10mg , Bafetinib CAS:859212-16-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY5086810MG
Encompass_Preferred
HY-50868 10mg Bafetinib CAS: 859212-16-1 Bafetinib is a potent and orally active Lyn/Bcr-Abl tyrosine kinase inhibitor. Bafetinib augments the activities of several proapoptotic Bcl-2 homology (BH)3-only proteins (Bim, Bad, Bmf and Bik) and induces apoptosis in Ph+ leukemia cells via Bcl-2 family-regulated intrinsic apoptosis pathway. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1083