Learn More
Selleck Chemical LLC Bafetinib (INNO-406) 5mg 859212-16-1 NS-187
SDP
Catalog No. 507661
Encompass_Preferred
Bafetinib (INNO-406, NS-187) is a potent and selective dual Bcr-Abl/Lyn inhibitor with IC50 of 5.8 nM/19 nM in cell-free assays, does not inhibit the phosphorylation of the T315I mutant and is less potent to PDGFR and c-Kit. Phase 2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-766-1 Supplier Selleck Chemical LLC Supplier No. S13695MG
Bafetinib (INNO-406, NS-187) is a potent and selective dual Bcr-Abl/Lyn inhibitor with IC50 of 5.8 nM/19 nM in cell-free assays, does not inhibit the phosphorylation of the T315I mutant and is less potent to PDGFR and c-Kit. Phase 2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.