Learn More
ACROBiosystems ACROBiosystems Biotinylated Human BAFF / TNFSF13B / CD257 Protein, Avitag™,Fc Tag (active trimer) (MALS verified)
Catalog No. 501957434
Encompass_Preferred
Recombinant Protein;200UG;Biotinylated Human BAFF, Avitag,Fc Tag (active trimer) (MALS verified) (BAF-H82F3) is expressed from human 293 cells (HEK293). It contains AA Ala 134 - Leu 285 (Accession # AAH20674.1).;This protein carries an Avi tag (Avitag™) at the N-terminus, followed by a human IgG1 Fc tag. The protein has a calculated MW of 44.9 kDa. The protein migrates as 50-55 kDa under reducing (R) condition (SDS-PAGE) due to Glycosylation.;Immobilized Human BCMA, Fc Tag (Cat. No. BC7-H5254) at 5 μg/mL (100 μL/well) can bind Biotinylated Human BAFF, Avitag,Fc Tag (active trimer) (MALS verified) (Cat. No. BAF-H82F3) with a linear range of 1.95-15.6 ng/mL (QC tested).
Catalog No. 50-195-7434 Supplier ACROBiosystems Supplier No. BAFH82F3200UG
Recombinant Protein;200UG;Biotinylated Human BAFF, Avitag,Fc Tag (active trimer) (MALS verified) (BAF-H82F3) is expressed from human 293 cells (HEK293). It contains AA Ala 134 - Leu 285 (Accession # AAH20674.1).;This protein carries an Avi tag (Avitag™) at the N-terminus, followed by a human IgG1 Fc tag. The protein has a calculated MW of 44.9 kDa. The protein migrates as 50-55 kDa under reducing (R) condition (SDS-PAGE) due to Glycosylation.;Immobilized Human BCMA, Fc Tag (Cat. No. BC7-H5254) at 5 μg/mL (100 μL/well) can bind Biotinylated Human BAFF, Avitag,Fc Tag (active trimer) (MALS verified) (Cat. No. BAF-H82F3) with a linear range of 1.95-15.6 ng/mL (QC tested).