Learn More
ACROBiosystems ACROBiosystems Biotinylated Human BAFF / TNFSF13B / CD257 Protein, His,Avitag™ (active trimer) (MALS verified)
Catalog No. 501957429
Encompass_Preferred
Recombinant Protein;200UG;Biotinylated Human BAFF, His,Avitag (active trimer) (MALS verified) (BAF-H82Q2) is expressed from human 293 cells (HEK293). It contains AA Ala 134 - Leu 285 (Accession # AAH20674.1).;This protein carries a polyhistidine tag at the N-terminus, followed by an Avi tag (Avitag™). The protein has a calculated MW of 20.2 kDa. The protein migrates as 21-23 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human BCMA, Fc Tag (Cat. No. BC7-H5254) at 2 μg/mL (100 μL/well) can bind Biotinylated Human BAFF, His,Avitag (active trimer) (MALS verified) (Cat. No. BAF-H82Q2) with a linear range of 0.2-4 ng/mL (QC tested).;Immobilized Human BCMA, His Tag (Cat. No. BCA-H522y) at 5 μg/mL (100 μL/well) can bind Biotinylated Human BAFF, His,Avitag (active trimer) (MALS verified) (Cat. No. BAF-H82Q2) with a linear range of 0.2-4 ng/mL (Routinely tested).
Catalog No. 50-195-7429 Supplier ACROBiosystems Supplier No. BAFH82Q2200UG
Recombinant Protein;200UG;Biotinylated Human BAFF, His,Avitag (active trimer) (MALS verified) (BAF-H82Q2) is expressed from human 293 cells (HEK293). It contains AA Ala 134 - Leu 285 (Accession # AAH20674.1).;This protein carries a polyhistidine tag at the N-terminus, followed by an Avi tag (Avitag™). The protein has a calculated MW of 20.2 kDa. The protein migrates as 21-23 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human BCMA, Fc Tag (Cat. No. BC7-H5254) at 2 μg/mL (100 μL/well) can bind Biotinylated Human BAFF, His,Avitag (active trimer) (MALS verified) (Cat. No. BAF-H82Q2) with a linear range of 0.2-4 ng/mL (QC tested).;Immobilized Human BCMA, His Tag (Cat. No. BCA-H522y) at 5 μg/mL (100 μL/well) can bind Biotinylated Human BAFF, His,Avitag (active trimer) (MALS verified) (Cat. No. BAF-H82Q2) with a linear range of 0.2-4 ng/mL (Routinely tested).