Learn More
Enzo Life Sciences BAFF (human) monoclonal antibody (Buffy-1) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX804128C100
Encompass_Preferred
Clone: Buffy-1. Isotype: IgG2a. Immunogen: Recombinant human soluble BAFF (aa 83-285) (ALX-522-025). Source: Purified from mouse ascites. Species reactivity: Human. Specificity: Recognizes membrane-bound BAFF. Detects endogenous protein by FC. Applications: Flow Cytometry, ICC, WB. Formulation: Liquid. In PBS containing 0.02% sodium azide.
Catalog No. 50-200-9788