Learn More
Enzo Life Sciences BAFF (mouse) monoclonal antibody (5A8) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX804158C100
Encompass_Preferred
Clone: 5A8. Isotype: IgG1. Immunogen: Rat RBL cells expressing mouse BAFF. Source: Purified from ascites. Species reactivity: Mouse. Applications: ELISA, Flow Cytometry, WB. Formulation: Liquid. In PBS containing 0.02% sodium azide.
Catalog No. 50-200-9805