Learn More
Enzo Life Sciences BAFF (mouse) monoclonal antibody (1C9) (biotin conjugate) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX804157BC100
Encompass_Preferred
Clone: 1C9. Isotype: IgG1. Immunogen: Rat RBL cells expressing mouse BAFF. Source: Purified from hybridoma tissue culture supernatant. Species reactivity: Mouse. Applications: ELISA, Flow Cytometry. Formulation: Liquid. In PBS containing 0.02% sodium azide.
Catalog No. 50-200-9803