Learn More
AdipoGen anti-BAFF (mouse), mAb (blocking) (Sandy-2) (100 µg)
SDP
Supplier:  AdipoGen AG20B0063C100
Encompass_Preferred
Monoclonal Antibody. Recognizes mouse BAFF. Isotype: Mouse IgG1. Clone: Sandy-2. Applications: FUNC (Blocking), IP. Liquid. In PBS containing 10% glycerol and 0.02% sodium azide. BAFF is a master regulator of peripheral B cell survival and, together with IL-6, promotes Ig class-switching and plasma cell differentiation. BAFF co-stimulates activated T cells. Increased levels of soluble BAFF have been detected in the serum of patients with various autoimmune diseases, such as Sjögren’s syndrome, rheumatoid arthritis, multiple sclerosis and systemic lupus erythematosus (SLE). Furthermore, BAFF is found in inflammatory sites in which there is lymphoid neogenesis. BAFF levels are elevated in patients with multiple myeloma and B cell chronic lymphoid leukemia (B-CCL).
Catalog No. 50-205-2976