Learn More
AdipoGen BAFF-R (human):Fc (human) (rec.) (50 µg)
SDP
Supplier:  AdipoGen AG40B0027C050
Encompass_Preferred
Protein. The extracellular domain of human BAFF-R (aa 2-71) is fused at the C-terminus to the Fc portion of human IgG1. Source: HEK 293 cells. Endotoxin content: <0.01EU/µg purified protein (LAL test; Lonza). Lyophilized. Contains PBS. Binds to human and mouse BAFF. Purity: >95% (SDS-PAGE). BAFF binds three distinct receptors (BAFF-R, TACI and BCMA) expressed predominantly on B cells, although activated T cells also express BAFF-R. BAFF is a master regulator of peripheral B cell survival, and together with IL-6, promotes Ig class-switching and plasma cell differentiation . Besides its major role in B cell biology, BAFF co-stimulates activated T cells. In humans, elevated levels of soluble BAFF have been detected in the serum of patients with various autoimmune diseases, such as Sjögren’s syndrome, Rheumatoid Arthritis (RA), Multiple sclerosis (MS) and Systemic Lupus Erythematosus (SLE) . BAFF is also increased levels in some lymphoid cancers.
Catalog No. 50-205-3100