Learn More
AdipoGen anti-BAFF-R (human), mAb (HuBR9.1) (Biotin) (100 µg)
SDP
Supplier:  AdipoGen AG20B0016BC100
Encompass_Preferred
Monoclonal Antibody. Recognizes human BAFF-R. Isotype: Mouse IgG1. Clone: HuBR9.1. Applications: FACS. Liquid. In PBS containing 0.02% sodium azide. Three receptors for BAFF were recently identified and designated TACI, BCMA and BAFF-R. BAFF-R signaling is essential for B cell development, survival and humoral immune responses.
Catalog No. 50-205-2911