Learn More
Enzo Life Sciences BAFF-R (human) : Fc (human), (recombinant) (50µg)
SDP
Catalog No. 502009398
Encompass_Preferred
Alternative name: TNFRSF 13C, BR3, BlySR3, CD268. Concentration: 1mg/ml (after reconstitution). Purity: ≥95% (SDS-PAGE). Formulation: Lyophilized. Contains PBS. Source: Produced in HEK 293 cells. The extracellular domain of human BAFF-R (aa 2-71) is fused at the C-terminus to the Fc portion of human IgG1. Specificity: Binds human and mouse BAFF. Long Term Storage: -20°C. Applications: ELISA .
Catalog No. 50-200-9398 Supplier Enzo Life Sciences Supplier No. ALX522060C050
Alternative name: TNFRSF 13C, BR3, BlySR3, CD268. Concentration: 1mg/ml (after reconstitution). Purity: ≥95% (SDS-PAGE). Formulation: Lyophilized. Contains PBS. Source: Produced in HEK 293 cells. The extracellular domain of human BAFF-R (aa 2-71) is fused at the C-terminus to the Fc portion of human IgG1. Specificity: Binds human and mouse BAFF. Long Term Storage: -20°C. Applications: ELISA .