Learn More
AdipoGen anti-BAFF-R (mouse), mAb (9B9) (100 µg)
SDP
Supplier:  AdipoGen AG20B0034C100
Encompass_Preferred
Monoclonal Antibody. Recognizes mouse BAFF-R. Isotype: Rat IgG2b. Clone: 9B9. Applications: FACS, FUNC. Liquid. In PBS containing 0.02% sodium azide. Three receptors for BAFF were recently identified and designated TACI, BCMA and BAFF-R. BAFF-R signaling is essential for B cell development, survival and humoral immune responses.
Catalog No. 50-205-2936