Learn More
AdipoGen BAFF, Soluble (human) Matched Pair Detection Set (1 Set)
SDP
Catalog No. 502053332
Encompass_Preferred
Matched Pair Detection Set. Detects human BAFF. Does not detect mouse BAFF. Colorimetric assay. Sample Types: Cell Culture Supernatant, Plasma, Serum. Range: 16 to 1000pg/ml. Sensitivity: 16pg/ml. BAFF (B cell-activating factor; BLyS; B lymphocyte stimulator; TNFSF13B; TALL-1; CD257), belonging to the TNF family, is a master regulator of peripheral B cell survival, and together with IL-6, promotes Ig class-switching and plasma cell differentiation. BAFF co-stimulates activated T cells. Increased levels of soluble BAFF have been detected in the serum of patients with various autoimmune diseases, such as Sjögren’s syndrome, rheumatoid arthritis, multiple sclerosis and systemic lupus erythematosus (SLE). Furthermore, BAFF is found in inflammatory sites in which there is lymphoid neogenesis. BAFF levels are elevated in patients with multiple myeloma and B cell chronic lymphoid leukemia (B-CCL).
Catalog No. 50-205-3332 Supplier AdipoGen Supplier No. AG46B0001KI01
Matched Pair Detection Set. Detects human BAFF. Does not detect mouse BAFF. Colorimetric assay. Sample Types: Cell Culture Supernatant, Plasma, Serum. Range: 16 to 1000pg/ml. Sensitivity: 16pg/ml. BAFF (B cell-activating factor; BLyS; B lymphocyte stimulator; TNFSF13B; TALL-1; CD257), belonging to the TNF family, is a master regulator of peripheral B cell survival, and together with IL-6, promotes Ig class-switching and plasma cell differentiation. BAFF co-stimulates activated T cells. Increased levels of soluble BAFF have been detected in the serum of patients with various autoimmune diseases, such as Sjögren’s syndrome, rheumatoid arthritis, multiple sclerosis and systemic lupus erythematosus (SLE). Furthermore, BAFF is found in inflammatory sites in which there is lymphoid neogenesis. BAFF levels are elevated in patients with multiple myeloma and B cell chronic lymphoid leukemia (B-CCL).