Learn More
AdipoGen BAFF, Soluble (mouse) (rec.) MultiPack,Protein
SDP
Supplier:  AdipoGen AG40B00223010
Encompass_Preferred
BAFF, Soluble (mouse) (rec.) MultiPack, AdipoGen Life Sciences. Protein. Mouse BAFF (aa 127-309) is fused at the N-terminus to a FLAG?-tag.  Lyophilized. Contains PBS.
Catalog No. 50-114-6065