Learn More
Enzo Life Sciences BAFF (soluble) (mouse), (recombinant) (10µg)
SDP
Supplier:  Enzo Life Sciences ALX522052C010
Encompass_Preferred
Alternative name: BLyS, TALL-1, THANK, zTNF4, TNFSF 13B/20, CD257. Concentration: 0.1mg/ml after reconstitution. Purity: ≥90% (SDS-PAGE). Formulation: Lyophilized. Contains PBS. Source: Produced in HEK 293 cells. The extracellular domain of mouse BAFF (aa 127-309) is fused at the N-terminus to a linker peptide (10 aa) and a FLAG-tag. Specificity: Binds to human (weak) and mouse BCMA, TACI and BAFF-R. Long Term Storage: -20°C. Applications: ELISA .
Catalog No. 50-200-9392